Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048168.2 | NP_001041633.1 | 3 - 4 | 327 | 94 | |
NM_008692.4 | NP_032718.2 | 3 - 4 | 400 | 94 | ||
GenBank mRNA | AB006181.1 | - | 2 - 3 | 180 | 94 | |
AK011327.1 | - | 3 - 4 | 357 | 94 | ||
BC020117.1 | - | 3 - 4 | 326 | 4009737 | 94 | |
BC053723.1 | - | 3 - 4 | 330 | 30053020 | 94 | |
BC085261.1 | - | 3 - 4 | 326 | 6397760 | 94 | |
CF172077.1 | - | 3 - 4 | 323 | 94 | ||
CJ092564.1 | - | 3 - 4 | 377 | 94 | ||
DV650959.1 | - | 3 - 4 | 339 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |