Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 61 - 62 | 10260 | 73 | |
NM_010862.2 | NP_034992.2 | 62 - 63 | 10314 | 73 | ||
NM_182698.2 | NP_874357.2 | 60 - 61 | 6498 | 73 | ||
GenBank mRNA | AF144095.1 | - | 62 - 63 | 10314 | 73 | |
AY331132.1 | - | 61 - 62 | 10260 | 73 | ||
AY331133.1 | - | 60 - 61 | 6498 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |