Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010439.3 | NP_034569.1 | 4 | 596 | 109 | |
GenBank mRNA | AK037928.1 | - | 4 | 631 | 109 | |
AK050659.1 | - | 4 | 601 | 109 | ||
AK051800.1 | - | 4 | 522 | 109 | ||
AK082902.1 | - | 4 | 578 | 109 | ||
AK084841.1 | - | 4 | 597 | 109 | ||
AK087949.1 | - | 4 | 565 | 109 | ||
AK134228.1 | - | 3 | 334 | 109 | ||
AK144428.1 | - | 572 | 109 | |||
AK146420.1 | - | 4 | 542 | 109 | ||
AK150924.1 | - | 4 | 600 | 109 | ||
AK157846.1 | - | 4 | 597 | 109 | ||
AK166007.1 | - | 4 | 551 | 109 | ||
BC006586.1 | - | 4 | 533 | 3587109 | 109 | |
BC008565.1 | - | 4 | 571 | 3586745 | 109 | |
BC064790.1 | - | 881 | 30019980 | 109 | ||
BC083067.1 | - | 4 | 519 | 5372544 | 109 | |
BC085090.1 | - | 4 | 552 | 6485915 | 109 | |
BC091741.1 | - | 4 | 556 | 4987137 | 109 | |
BC094030.1 | - | 4 | 586 | 4952464 | 109 | |
BC100312.1 | - | 4 | 574 | 30849071 | 109 | |
BC110667.1 | - | 3 | 1008 | 5036554 | 109 | |
BC111468.1 | - | 4 | 522 | 4215684 | 109 | |
L38477.1 | - | 4 | 517 | 109 | ||
U00431.1 | - | 4 | 589 | 109 | ||
Z11997.1 | - | 4 | 516 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Amplicon spans exons and probe does not span exons |