Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200004708.mm.1 |
NM_007494.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004708.mm.1 |
NM_007494.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004708.mm.1 |
TC0200000551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004711.mm.1 |
NM_007494.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004711.mm.1 |
NM_007494.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004711.mm.1 |
NR_002687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004711.mm.1 |
TC0200000551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004713.mm.1 |
NM_007494.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004713.mm.1 |
NM_007494.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004713.mm.1 |
NR_002687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004713.mm.1 |
TC0200000551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004712.mm.1 |
NM_007494.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004712.mm.1 |
NM_007494.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004712.mm.1 |
NR_002687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004712.mm.1 |
TC0200000551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004709.mm.1 |
NM_007494.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004709.mm.1 |
NM_007494.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004709.mm.1 |
NR_002687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004709.mm.1 |
TC0200000551.mm.1 |
MTA TC |