Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026614.2 | NP_080890.1 | 1 - 2 | 167 | 66 | |
GenBank mRNA | AK009338.1 | - | 1 - 2 | 51 | 66 | |
AK009642.1 | - | 1 - 2 | 52 | 66 | ||
AK009775.1 | - | 1 - 2 | 61 | 66 | ||
AK009840.1 | - | 1 - 2 | 51 | 66 | ||
AK009985.1 | - | 1 - 2 | 51 | 66 | ||
AK010092.1 | - | 1 - 2 | 53 | 66 | ||
AK019245.1 | - | 1 - 2 | 58 | 66 | ||
AK019301.1 | - | 1 - 2 | 168 | 66 | ||
AK019324.1 | - | 1 - 2 | 147 | 66 | ||
AK019359.1 | - | 1 - 2 | 59 | 66 | ||
AK019364.1 | - | 1 - 2 | 53 | 66 | ||
AK019368.1 | - | 1 - 2 | 51 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |