Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028770.2 | NP_083046.2 | 6 - 7 | 1030 | 107 | |
GenBank mRNA | AK004811.2 | - | 5 - 6 | 819 | 107 | |
AK171114.1 | - | 919 | 107 | |||
BC119366.1 | - | 6 - 7 | 1030 | 8734115 | 107 | |
BC120601.1 | - | 6 - 7 | 1030 | 40129524 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |