Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
NM_001271483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
NM_001271484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
NM_030179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
NM_030179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
NM_175378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
NM_175378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
NR_073185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010785.mm.1 |
TC1700001143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010782.mm.1 |
NM_030179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010782.mm.1 |
NM_030179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010782.mm.1 |
TC1700001143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
NM_001271483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
NM_001271484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
NM_030179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
NM_030179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
NM_175378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
NM_175378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
NR_073185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010787.mm.1 |
TC1700001143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
NM_001271483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
NM_001271484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
NM_030179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
NM_030179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
NM_175378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
NM_175378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
NR_073185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010784.mm.1 |
TC1700001143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
NM_001271483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
NM_001271484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
NM_030179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
NM_030179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
NM_175378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
NM_175378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
NR_073185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010786.mm.1 |
TC1700001143.mm.1 |
MTA TC |