Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172624.3 | NP_766212.2 | 13 - 14 | 1737 | 77 | |
GenBank mRNA | AK050021.1 | - | 13 - 14 | 1679 | 77 | |
AK050023.1 | - | 13 - 14 | 1739 | 77 | ||
AK078301.1 | - | 13 - 14 | 1685 | 77 | ||
AK131178.1 | - | 10 - 11 | 3600 | 77 | ||
AK204395.1 | - | 2 - 3 | 242 | 77 | ||
BC057631.1 | - | 13 - 14 | 1736 | 4191886 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |