Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177356.3 | NP_796330.2 | 3 - 4 | 974 | 91 | |
GenBank mRNA | AJ510130.1 | - | 3 - 4 | 893 | 91 | |
AK028039.1 | - | 3 - 4 | 926 | 91 | ||
AK033379.1 | - | 3 - 4 | 927 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |