Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029733.1 | 1 | 43 | 78 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
deleted in lymphocytic leukemia, 2 |
Dleu2 |
1810047A16Rik, 4932411A06Rik, 7730401J12Rik, AI197277, AI788824, Alt1, Gm9069, Leu2 |
Sense | Chr. 14 - 61602836 - 61682373 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 27017 |
Gm27017 | 47.9kb | Anti-sense | Chr. 14 - 61680039 - 61686642 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |