Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242615.2 | NP_001229544.1 | 7 - 8 | 843 | 93 | |
NM_001242616.2 | NP_001229545.1 | 7 - 8 | 840 | 93 | ||
NM_001290738.1 | NP_001277667.1 | 6 - 7 | 762 | 93 | ||
GenBank mRNA | AK160327.1 | - | 7 - 8 | 809 | 93 | |
AY231460.1 | - | 7 - 8 | 559 | 93 | ||
AY258040.1 | - | 7 - 8 | 559 | 93 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | deoxyribonuclease 1-like 1 | Dnase1l1 |
2310005K03Rik, AI324885, Dnase1ll, Dnasex, Dnl1ll, G4.8 |
-6.1kb | Anti-sense | Chr. X - 74273217 - 74282333 on NCBI Build GRCm38 |
Downstream |
ATPase, H+ transporting, lysosomal accessory protein 1 |
Atp6ap1 |
16A, AC45, AI316502, AW108110, Atp6ip1, Atp6s1, C7-1, CF2, VATPS1, XAP-3, mFLJ00383 |
7.9kb | Sense | Chr. X - 74297097 - 74304721 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |