Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204333.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204333.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_001204336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_022434.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
NM_022434.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017160.mm.1 |
TC1700001856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204333.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204333.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_001204336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_022434.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
NM_022434.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017159.mm.1 |
TC1700001856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204333.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204333.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_001204336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_022434.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
NM_022434.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017158.mm.1 |
TC1700001856.mm.1 |
MTA TC |