Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111937.1 | 1 - 2 | 124 | 53 | ||
GenBank mRNA | AK152931.1 | - | 1 - 2 | 59 | 53 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nucleoporin 98 | Nup98 |
4732457F17, AI849286, Nup96 |
-12.9kb | Anti-sense | Chr. 7 - 102119400 - 102210166 on NCBI Build GRCm38 |
Downstream |
ras homolog gene family, member G |
Rhog |
2810426G09Rik, Arhg, Sid10750 |
12.7kb | Anti-sense | Chr. 7 - 102239123 - 102250118 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |