Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289455.1 | NP_001276384.1 | 2 - 3 | 300 | 60 | |
GenBank mRNA | BC104732.1 | - | 2 - 3 | 264 | 40046865 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |