Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009969.4 | NP_034099.2 | 2 - 3 | 481 | 70 | |
GenBank mRNA | AK053196.1 | - | 2 - 3 | 481 | 70 | |
BC116878.1 | - | 2 - 3 | 215 | 40126197 | 70 | |
BC116880.1 | - | 2 - 3 | 215 | 40126199 | 70 | |
EU366957.1 | - | 2 - 3 | 192 | 70 | ||
M11848.1 | - | 2 - 3 | 150 | 70 | ||
X02333.1 | - | 2 - 3 | 365 | 70 | ||
X03019.1 | - | 2 - 3 | 224 | 70 | ||
X03221.1 | - | 2 - 3 | 223 | 70 | ||
X05906.1 | - | 2 - 3 | 123 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |