Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103661.3 | XP_011959051.1 | 6 - 7 | 790 | 110 | |
XM_027960930.1 | XP_027816731.1 | 6 - 7 | 830 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |