Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009778.1 | NP_001009778.1 | 5 - 6 | 552 | 90 | |
GenBank mRNA | U12761.1 | - | 5 - 6 | 552 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |