Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009736.1 | NP_001009736.1 | 4 - 5 | 324 | 92 | |
XM_012146332.3 | XP_012001722.2 | 3 - 4 | 434 | 92 | ||
GenBank mRNA | AF173557.1 | - | 4 - 5 | 324 | 92 |