Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009415.1 | NP_001009415.1 | 1 - 2 | 303 | 67 | |
GenBank mRNA | X60149.1 | - | 164 | 67 | ||
Z11560.1 | - | 303 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |