Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011704.4 | XP_004011753.2 | 2 - 3 | 412 | 96 | |
XM_012183770.2 | XP_012039160.2 | 1 - 2 | 68 | 96 |