Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012640.3 | XP_004012689.3 | 3 - 4 | 425 | 92 | |
XM_027974997.1 | XP_027830798.1 | 3 - 4 | 150 | 92 |