Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012640.3 | XP_004012689.3 | 4 - 5 | 1250 | 71 | |
XM_027974997.1 | XP_027830798.1 | 4 - 5 | 975 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |