Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126352.1 | NP_001119824.1 | 1 | 317 | 71 | |
XM_015095469.2 | XP_014950955.1 | 5 - 6 | 430 | 71 | ||
XM_015095470.2 | XP_014950956.1 | 5 - 6 | 411 | 71 | ||
XM_027969158.1 | XP_027824959.1 | 5 - 6 | 403 | 71 | ||
GenBank mRNA | EU596596.1 | - | 1 | 317 | 71 |
Gene Symbol: LOC114116592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973508.1 | XP_027829309.1 | 1 | 342 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |