Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166184.1 | NP_001159656.1 | 4 - 5 | 732 | 66 | |
GenBank mRNA | DQ152963.1 | - | 1 - 2 | 66 | 66 | |
EU366482.1 | - | 1 - 2 | 94 | 66 | ||
GQ221037.1 | - | 4 - 5 | 732 | 66 |