Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126354.1 | NP_001119826.1 | 1 - 2 | 137 | 63 | |
GenBank mRNA | EU596595.1 | - | 1 - 2 | 137 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |