Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013753.3 | XP_004013802.1 | 1 - 2 | 133 | 55 | |
XM_012187380.3 | XP_012042770.2 | 1 - 2 | 122 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |