Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184538.3 | XP_012039928.1 | 2 - 3 | 678 | 53 | |
XM_015098094.2 | XP_014953580.1 | 2 - 3 | 701 | 53 | ||
XM_027973598.1 | XP_027829399.1 | 2 - 3 | 878 | 53 | ||
XM_027973599.1 | XP_027829400.1 | 2 - 3 | 878 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |