Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186426.3 | XP_012041816.2 | 2 - 3 | 3313 | 86 | |
XM_015098909.2 | XP_014954395.2 | 2 - 3 | 276 | 86 | ||
XM_027974284.1 | XP_027830085.1 | 1 - 2 | 203 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |