Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004565.3 | XP_004004614.2 | 1 - 2 | 866 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |