Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008736.4 | XP_004008785.2 | 3 - 4 | 1202 | 64 | |
XM_012178352.3 | XP_012033742.2 | 3 - 4 | 1206 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |