Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163057.1 | NP_001156529.1 | 1 - 2 | 175 | 70 | |
GenBank mRNA | FJ969407.1 | - | 1 - 2 | 175 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |