Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095891.2 | XP_014951377.2 | 1 - 2 | 803 | 86 | |
XM_015095893.2 | XP_014951379.2 | 2 - 3 | 444 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |