Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002953.4 | XP_004003002.2 | 1 - 2 | 591 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |