Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975461.1 | XP_027831262.1 | 2 - 3 | 239 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |