Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093551.2 | XP_014949037.2 | 2 - 3 | 576 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |