Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001772.4 | XP_004001821.1 | 1 - 2 | 526 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |