Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009304.1 | NP_001009304.1 | 2 - 3 | 681 | 71 | |
XM_027956983.1 | XP_027812784.1 | 2 - 3 | 670 | 71 | ||
GenBank mRNA | AY679518.1 | - | 1 - 2 | 633 | 71 | |
M21027.1 | - | 2 - 3 | 681 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |