Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114767.2 | NP_001108239.1 | 1 - 2 | 332 | 67 | |
GenBank mRNA | KT853038.1 | - | 1 - 2 | 357 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |