Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136491.1 | NP_001129963.1 | 5 - 6 | 776 | 107 | |
GenBank mRNA | FJ211194.1 | - | 5 - 6 | 776 | 107 |
Gene Symbol: LOC114118377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979143.1 | XP_027834944.1 | 5 - 6 | 837 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |