Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123403.1 | NP_001116875.1 | 4 - 5 | 894 | 100 | |
XM_027977361.1 | XP_027833162.1 | 5 - 6 | 968 | 100 | ||
GenBank mRNA | AF421499.1 | - | 4 - 5 | 894 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |