Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114763.2 | NP_001108235.1 | 5 - 6 | 1021 | 113 | |
GenBank mRNA | EU216424.1 | - | 5 - 6 | 1021 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |