Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009274.1 | NP_001009274.1 | 2 - 3 | 324 | 88 | |
XM_027967511.1 | XP_027823312.1 | 2 - 3 | 517 | 88 | ||
GenBank mRNA | AY205232.1 | - | 2 - 3 | 324 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |