Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024963.2 | NP_001020134.1 | 5 - 6 | 1102 | 91 | |
XM_012174077.3 | XP_012029467.1 | 5 - 6 | 1344 | 91 | ||
GenBank mRNA | AY390555.1 | - | 5 - 6 | 1105 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |