Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093780.1 | NP_001087249.1 | 9 - 10 | 1154 | 82 | |
XM_027962764.1 | XP_027818565.1 | 10 - 11 | 1311 | 82 | ||
XM_027962765.1 | XP_027818566.1 | 10 - 11 | 1281 | 82 | ||
XM_027962766.1 | XP_027818567.1 | 9 - 10 | 1575 | 82 | ||
XM_027962767.1 | XP_027818568.1 | 9 - 10 | 1279 | 82 | ||
XM_027962768.1 | XP_027818569.1 | 9 - 10 | 1203 | 82 | ||
XM_027962769.1 | XP_027818570.1 | 9 - 10 | 1173 | 82 | ||
GenBank mRNA | DQ377363.1 | - | 9 - 10 | 1124 | 82 | |
DQ377364.1 | - | 9 - 10 | 1154 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |