Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075101.1 | NP_001068569.1 | 4 - 5 | 406 | 74 | |
GenBank mRNA | DQ054533.1 | - | 4 - 5 | 406 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |