Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127271.1 | NP_001120743.1 | 1 - 2 | 59 | 96 | |
GenBank mRNA | EU626216.1 | - | 1 - 2 | 59 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |