Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267884.2 | NP_001254813.1 | 1 - 2 | 182 | 96 | |
XM_012110711.2 | XP_011966101.1 | 2 - 3 | 96 | |||
GenBank mRNA | JQ780322.1 | - | 1 - 2 | 186 | 96 |