Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973466.1 | XP_027829267.1 | 4 - 5 | 1179 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |