Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958558.1 | XP_027814359.1 | 1 - 2 | 97 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |