Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012188597.3 | XP_012043987.1 | 3 - 4 | 455 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |